siRNA Details
| Gene Symbol: LOC283856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027078.1 | 1 | 656 | |||
| GenBank mRNA | BC035113.2 | - | 1 | 656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283856| Gene Symbol: LOC283856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027078.1 | 1 | 656 | |||
| GenBank mRNA | BC035113.2 | - | 1 | 656 | ||