siRNA Details
| Gene Symbol: MDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027042.1 | 4 | 536 | |||
| GenBank mRNA | AJ310434.2 | - | 4 | 536 | ||
| BC041472.1 | - | 5 | 1431 | |||
| BC069164.1 | - | 4 | 536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MDS2| Gene Symbol: MDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027042.1 | 4 | 536 | |||
| GenBank mRNA | AJ310434.2 | - | 4 | 536 | ||
| BC041472.1 | - | 5 | 1431 | |||
| BC069164.1 | - | 4 | 536 | |||