siRNA Details
| Gene Symbol: MDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027042.1 | 2 | 244 | |||
| GenBank mRNA | AJ310434.2 | - | 2 | 244 | ||
| BC069164.1 | - | 2 | 244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MDS2| Gene Symbol: MDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027042.1 | 2 | 244 | |||
| GenBank mRNA | AJ310434.2 | - | 2 | 244 | ||
| BC069164.1 | - | 2 | 244 | |||