siRNA Details
| Gene Symbol: LINC00922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027755.1 | 3 | 341 | |||
| GenBank mRNA | BC037902.1 | - | 3 | 347 | ||
| BC104446.1 | - | 5 | 472 | |||
| BC104447.1 | - | 2 | 143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENSG00000261742
LINC00922| Gene Symbol: LINC00922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027755.1 | 3 | 341 | |||
| GenBank mRNA | BC037902.1 | - | 3 | 347 | ||
| BC104446.1 | - | 5 | 472 | |||
| BC104447.1 | - | 2 | 143 | |||