siRNA Details
| Gene Symbol: MYH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002147.2 | 9 | 1240 | |||
| GenBank mRNA | AK025690.1 | - | 9 | 1240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYH16| Gene Symbol: MYH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002147.2 | 9 | 1240 | |||
| GenBank mRNA | AK025690.1 | - | 9 | 1240 | ||