siRNA Details
| Gene Symbol: ANKRD30BL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027019.1 | 7 | 1540 | |||
| NR_027020.2 | 5 | 1238 | ||||
| GenBank mRNA | BC045801.1 | - | 5 | 1238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD30BL| Gene Symbol: ANKRD30BL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027019.1 | 7 | 1540 | |||
| NR_027020.2 | 5 | 1238 | ||||
| GenBank mRNA | BC045801.1 | - | 5 | 1238 | ||