siRNA Details
| Gene Symbol: PMCHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003922.1 | 2 | 384 | |||
| GenBank mRNA | AY008412.1 | - | 2 | 384 | ||
| AY008413.1 | - | 2 | 384 | |||
| Gene Symbol: PMCHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY008411.1 | - | 2 | 384 | ||

PMCHL2
Genomic Map