siRNA Details
| Gene Symbol: LINC00626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024160.1 | 1 | 23 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00626| Gene Symbol: LINC00626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024160.1 | 1 | 23 | |||