siRNA Details
| Gene Symbol: MEG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024149.2 | 3 | 419 | |||
| GenBank mRNA | CA396130.1 | - | 3 | 422 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | T85042.1 | - | 329 | Chr. - - [-] on Build | ||

MEG8
Genomic Map