siRNA Details
| Gene Symbol: LINC01566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027079.2 | 8 | 1894 | |||
| NR_027080.2 | 6 | 939 | ||||
| GenBank mRNA | BC038774.1 | - | 6 | 939 | ||
| BC045579.1 | - | 8 | 1894 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01566| Gene Symbol: LINC01566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027079.2 | 8 | 1894 | |||
| NR_027080.2 | 6 | 939 | ||||
| GenBank mRNA | BC038774.1 | - | 6 | 939 | ||
| BC045579.1 | - | 8 | 1894 | |||