siRNA Details
| Gene Symbol: LINC01566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027079.2 | 4 | 709 | |||
| NR_027080.2 | 4 | 522 | ||||
| GenBank mRNA | BC038774.1 | - | 4 | 522 | ||
| BC045579.1 | - | 4 | 709 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI464475.1 | - | 4 | 527 | Chr. 16 - 35363412 - 35382466 [+] on Build GRCh38 | |

LINC01566
Genomic Map