siRNA Details
| Gene Symbol: FAM193B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024019.3 | 7 | 2616 | |||
| XM_011534582.1 | XP_011532884.1 | 6 | 2056 | |||
| XM_011534583.1 | XP_011532885.1 | 6 | 1797 | |||
| GenBank mRNA | AB067518.1 | - | 7 | 2573 | ||
| AK096032.1 | - | 5 | 768 | |||
| BC086859.1 | - | 3 | 430 | |||
| BX647587.1 | - | 4 | 701 | |||

FAM193B
Genomic Map