siRNA Details
| Gene Symbol: LOC283922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026950.1 | 15 | 2272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283922| Gene Symbol: LOC283922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026950.1 | 15 | 2272 | |||