siRNA Details
| Gene Symbol: FBXL19-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024348.1 | 1 | 329 | |||
| GenBank mRNA | AK092263.1 | - | 1 | 329 | ||
| BC101464.1 | - | 1 | 255 | |||
| BC101988.1 | - | 1 | 255 | |||
| BC105687.1 | - | 1 | 255 | |||
| BC105926.1 | - | 1 | 255 | |||

FBXL19-AS1
Genomic Map