siRNA Details
| Gene Symbol: TMEM185B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024121.2 | NP_077026.2 | 1 | 1617 | ||
| GenBank mRNA | AF530474.1 | - | 1 | 1617 | ||
| AK289880.1 | - | 1 | 1579 | |||
| BC080607.1 | - | 1 | 1584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM185B| Gene Symbol: TMEM185B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024121.2 | NP_077026.2 | 1 | 1617 | ||
| GenBank mRNA | AF530474.1 | - | 1 | 1617 | ||
| AK289880.1 | - | 1 | 1579 | |||
| BC080607.1 | - | 1 | 1584 | |||