siRNA Details
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027696.1 | 3 | 1221 | |||
NR_027697.1 | 3 | 1170 | ||||
GenBank mRNA | DA797317.1 | - | 1 | 486 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027696.1 | 3 | 1221 | |||
NR_027697.1 | 3 | 1170 | ||||
GenBank mRNA | DA797317.1 | - | 1 | 486 |