siRNA Details
| Gene Symbol: SCAND2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003654.2 | 4 | 1373 | |||
| NR_004859.1 | 5 | 1931 | ||||
| GenBank mRNA | AK022844.1 | - | 4 | 1373 | ||
| BC011547.1 | - | 4 | 1355 | |||
| BC012929.1 | - | 4 | 1355 | |||
| BC063836.1 | - | 4 | 1329 | |||

SCAND2P
Genomic Map