siRNA Details
Gene Symbol: SCAND2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003654.2 | 4 | 3508 | |||
NR_004859.1 | 5 | 4066 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SCAND2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003654.2 | 4 | 3508 | |||
NR_004859.1 | 5 | 4066 |