siRNA Details
| Gene Symbol: LINC00685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027231.1 | 2 | 743 | |||
| NR_027232.1 | 1 | 1399 | ||||
| GenBank mRNA | AK023454.1 | - | 1 | 1358 | ||
| BC041934.1 | - | 2 | 746 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00685| Gene Symbol: LINC00685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027231.1 | 2 | 743 | |||
| NR_027232.1 | 1 | 1399 | ||||
| GenBank mRNA | AK023454.1 | - | 1 | 1358 | ||
| BC041934.1 | - | 2 | 746 | |||