siRNA Details
| Gene Symbol: LINC00469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027146.1 | 4 | 1117 | |||
| GenBank mRNA | AK097638.1 | - | 4 | 1235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00469| Gene Symbol: LINC00469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027146.1 | 4 | 1117 | |||
| GenBank mRNA | AK097638.1 | - | 4 | 1235 | ||