siRNA Details
| Gene Symbol: CNTFR-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024369.1 | 2 | 607 | |||
| GenBank mRNA | BC033987.1 | - | 3 | 718 | ||
| BC038740.1 | - | 5 | 1040 | |||
| BC042806.1 | - | 2 | 608 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNTFR-AS1| Gene Symbol: CNTFR-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024369.1 | 2 | 607 | |||
| GenBank mRNA | BC033987.1 | - | 3 | 718 | ||
| BC038740.1 | - | 5 | 1040 | |||
| BC042806.1 | - | 2 | 608 | |||