siRNA Details
| Gene Symbol: FAM83A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024479.1 | 3 | 1001 | |||
| GenBank mRNA | AY211906.1 | - | 2 | 662 | ||
| AY211907.1 | - | 3 | 1001 | |||
| AY211908.1 | - | 2 | 1190 | |||
| AY225521.1 | - | 1 | 1415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM83A-AS1| Gene Symbol: FAM83A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024479.1 | 3 | 1001 | |||
| GenBank mRNA | AY211906.1 | - | 2 | 662 | ||
| AY211907.1 | - | 3 | 1001 | |||
| AY211908.1 | - | 2 | 1190 | |||
| AY225521.1 | - | 1 | 1415 | |||