siRNA Details
| Gene Symbol: JPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024582.1 | 5 | 976 | |||
| GenBank mRNA | BC071776.1 | - | 5 | 972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

JPX| Gene Symbol: JPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024582.1 | 5 | 976 | |||
| GenBank mRNA | BC071776.1 | - | 5 | 972 | ||