siRNA Details
| Gene Symbol: TMEM30C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028357.1 | 2 | 92 | |||
| NR_028358.1 | 2 | 92 | ||||
| GenBank mRNA | AB249666.1 | - | 2 | 92 | ||
| AB250297.1 | - | 2 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM30C| Gene Symbol: TMEM30C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028357.1 | 2 | 92 | |||
| NR_028358.1 | 2 | 92 | ||||
| GenBank mRNA | AB249666.1 | - | 2 | 92 | ||
| AB250297.1 | - | 2 | 92 | |||