siRNA Details
| Gene Symbol: LINC00934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024246.2 | 4 | 1764 | |||
| GenBank mRNA | AK057497.1 | - | 4 | 1764 | ||
| BC101457.1 | - | 1 | 767 | |||
| BC101458.1 | - | 1 | 767 | |||
| BC101459.1 | - | 1 | 767 | |||
| BC118554.1 | - | 1 | 767 | |||
| BC118632.1 | - | 2 | 681 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000630357.1 | - | 1764 | Chr. - - [-] on Build | ||

LINC00934
Genomic Map