siRNA Details
Gene Symbol: LOC284009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028335.1 | 3 | 546 | |||
GenBank mRNA | BC093711.1 | - | 3 | 547 | 7939556 | |
BC112034.1 | - | 3 | 547 | 8327502 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028335.1 | 3 | 546 | |||
GenBank mRNA | BC093711.1 | - | 3 | 547 | 7939556 | |
BC112034.1 | - | 3 | 547 | 8327502 |