siRNA Details
| Gene Symbol: LINC00324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026951.1 | 2 | 749 | |||
| GenBank mRNA | AK092109.1 | - | 2 | 749 | ||
| BC063002.1 | - | 1 | 307 | |||
| BC089396.1 | - | 1 | 278 | |||
| BC113564.1 | - | 2 | 590 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00324| Gene Symbol: LINC00324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026951.1 | 2 | 749 | |||
| GenBank mRNA | AK092109.1 | - | 2 | 749 | ||
| BC063002.1 | - | 1 | 307 | |||
| BC089396.1 | - | 1 | 278 | |||
| BC113564.1 | - | 2 | 590 | |||