siRNA Details
| Gene Symbol: MIR137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029679.1 | 1 | 83 | |||
| Gene Symbol: MIR137HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046105.1 | 3 | 733 | |||
| GenBank mRNA | AK094607.1 | - | 3 | 645 | ||
| AK309618.1 | - | 1 | 133 | |||
| AK311400.1 | - | 1 | 136 | |||

MIR137
Genomic Map