siRNA Details
| Gene Symbol: HNRNPA1P33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003277.1 | 1 | 496 | |||
| GenBank mRNA | BC061889.1 | - | 1 | 297 | ||
| BC112994.1 | - | 1 | 496 | |||
| BC112995.1 | - | 1 | 496 | |||
| BC128038.1 | - | 1 | 496 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG742992.1 | - | 1 | 661 | Chr. 10 - 46415751 - 46416447 [+] on Build GRCh38 | |

HNRNPA1P33
Genomic Map