siRNA Details
| Gene Symbol: MIR187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029616.1 | 1 | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR187| Gene Symbol: MIR187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029616.1 | 1 | 14 | |||