siRNA Details
| Gene Symbol: BTN2A3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027795.1 | 3 | 1132 | |||
| GenBank mRNA | AK056871.1 | - | 3 | 1112 | ||
| BC143636.1 | - | 3 | 767 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BTN2A3P| Gene Symbol: BTN2A3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027795.1 | 3 | 1132 | |||
| GenBank mRNA | AK056871.1 | - | 3 | 1112 | ||
| BC143636.1 | - | 3 | 767 | |||