siRNA Details
| Gene Symbol: TMPO-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027157.1 | 2 | 2036 | |||
| GenBank mRNA | AK054586.1 | - | 1 | 668 | ||
| BC037346.2 | - | 1 | 1377 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ938655.1 | - | 176 | Chr. - - [-] on Build | ||

TMPO-AS1
Genomic Map