siRNA Details
| Gene Symbol: LOC202181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026921.1 | 6 | 2240 | |||
| GenBank mRNA | AK021623.1 | - | 1 | 623 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DN992215.1 | - | 4 | 504 | Chr. 5 - 177619059 - 177627500 [-] on Build GRCh38 | |

LOC202181
Genomic Map