siRNA Details
| Gene Symbol: MIR204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029621.1 | 1 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR204| Gene Symbol: MIR204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029621.1 | 1 | 95 | |||