siRNA Details
| Gene Symbol: MIR223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029637.1 | 1 | 96 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV712697.1 | - | 2 | 182 | Chr. X - 66017150 - 66019427 [+] on Build GRCh38 | |

MIR223
Genomic Map