siRNA Details
| Gene Symbol: FAM86JP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024250.1 | 5 | 738 | |||
| NR_024251.1 | 5 | 836 | ||||
| GenBank mRNA | BC032772.1 | - | 5 | 724 | ||
| BC041080.1 | - | 5 | 807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM86JP| Gene Symbol: FAM86JP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024250.1 | 5 | 738 | |||
| NR_024251.1 | 5 | 836 | ||||
| GenBank mRNA | BC032772.1 | - | 5 | 724 | ||
| BC041080.1 | - | 5 | 807 | |||