siRNA Details
| Gene Symbol: INTS4P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057766.1 | - | 8 | 1096 | ||
| Gene Symbol: INTS4P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027392.2 | 8 | 1103 | |||
| GenBank mRNA | BC111554.1 | - | 8 | 1096 | 40024359 | |

INTS4P1
Genomic Map