siRNA Details
| Gene Symbol: MIR98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029513.1 | 1 | 74 | |||
| GenBank mRNA | AF480570.1 | - | 1 | 54 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA362525.1 | - | 1 | 83 | Chr. X - 53556072 - 53556350 [-] on Build GRCh38 | |

MIR98
Genomic Map