siRNA Details
| Gene Symbol: ZNF733P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003952.1 | 1 | 34 | |||
| Gene Symbol: ZNF479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012777.1 | XP_016868266.1 | 1 | 72 | ||

ZNF733P
Genomic Map