siRNA Details
| Gene Symbol: LINC00314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027246.1 | 1 | 57 | |||
| GenBank mRNA | AF427489.1 | - | 1 | 38 | ||
| BC126226.1 | - | 1 | 14 | |||
| BC126230.1 | - | 1 | 14 | |||
| CB049373.1 | - | 1 | 57 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00314| Gene Symbol: LINC00314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027246.1 | 1 | 57 | |||
| GenBank mRNA | AF427489.1 | - | 1 | 38 | ||
| BC126226.1 | - | 1 | 14 | |||
| BC126230.1 | - | 1 | 14 | |||
| CB049373.1 | - | 1 | 57 | |||