siRNA Details
Gene Symbol: APOOP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028471.1 | 1 | 73 | |||
GenBank mRNA | BX108667.1 | - | 1 | 278 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: APOOP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028471.1 | 1 | 73 | |||
GenBank mRNA | BX108667.1 | - | 1 | 278 |