siRNA Details
| Gene Symbol: APOOP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028471.1 | 1 | 240 | |||
| GenBank mRNA | BX108667.1 | - | 1 | 445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

APOOP5| Gene Symbol: APOOP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028471.1 | 1 | 240 | |||
| GenBank mRNA | BX108667.1 | - | 1 | 445 | ||