siRNA Details
| Gene Symbol: OR3A4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024128.1 | 1 | 411 | |||
| GenBank mRNA | BC136874.1 | - | 1 | 411 | ||
| BC136875.1 | - | 1 | 411 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR3A4P| Gene Symbol: OR3A4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024128.1 | 1 | 411 | |||
| GenBank mRNA | BC136874.1 | - | 1 | 411 | ||
| BC136875.1 | - | 1 | 411 | |||