siRNA Details
| Gene Symbol: DLEU2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002771.1 | 1 | 274 | |||
| GenBank mRNA | AF254117.1 | - | 1 | 274 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022419.1 | - | 1 | 2172 | Chr. 1 - 63547082 - 63549558 [+] on Build GRCh38 | |

DLEU2L
Genomic Map