siRNA Details
| Gene Symbol: THAP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027051.1 | 1 | 659 | |||
| NR_027052.1 | 1 | 659 | ||||
| GenBank mRNA | AK096901.1 | - | 1 | 364 | ||
| BC110368.1 | - | 1 | 672 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU858779.1 | - | 1 | 23 | Chr. 22 - 21002558 - 21003265 [+] on Build GRCh38 | |

THAP7-AS1
Genomic Map