siRNA Details
Gene Symbol: THAP7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027051.1 | 1 | 571 | |||
NR_027052.1 | 1 | 571 | ||||
GenBank mRNA | AK096901.1 | - | 1 | 276 | ||
BC110368.1 | - | 1 | 584 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: THAP7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027051.1 | 1 | 571 | |||
NR_027052.1 | 1 | 571 | ||||
GenBank mRNA | AK096901.1 | - | 1 | 276 | ||
BC110368.1 | - | 1 | 584 |