siRNA Details
| Gene Symbol: LINC00575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024087.1 | 1 | 18 | |||
| NR_024088.1 | 1 | 18 | ||||
| GenBank mRNA | AY316301.1 | - | 1 | 18 | ||
| AY316302.1 | - | 1 | 18 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00575| Gene Symbol: LINC00575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024087.1 | 1 | 18 | |||
| NR_024088.1 | 1 | 18 | ||||
| GenBank mRNA | AY316301.1 | - | 1 | 18 | ||
| AY316302.1 | - | 1 | 18 | |||