siRNA Details
| Gene Symbol: OACYLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024021.2 | 4 | 512 | |||
| GenBank mRNA | DA088392.1 | - | 2 | 158 | ||
| DV080257.1 | - | 4 | 452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OACYLP| Gene Symbol: OACYLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024021.2 | 4 | 512 | |||
| GenBank mRNA | DA088392.1 | - | 2 | 158 | ||
| DV080257.1 | - | 4 | 452 | |||