siRNA Details
| Gene Symbol: OACYLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024021.2 | 2 | 251 | |||
| GenBank mRNA | DV080257.1 | - | 2 | 191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OACYLP| Gene Symbol: OACYLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024021.2 | 2 | 251 | |||
| GenBank mRNA | DV080257.1 | - | 2 | 191 | ||