siRNA Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015012.3 | NP_055827.1 | 7 | 3954 | ||
| NR_028491.2 | 8 | 1625 | ||||
| GenBank mRNA | BC035034.1 | - | 7 | 3832 | 5259179 | |
| BC044597.1 | - | 7 | 3822 | 4811565 | ||
| BC091524.1 | - | 8 | 1424 | 30512350 | ||

TMEM41B
Genomic Map